Thermo Scientific Chemicals

1,12-Diaminododecane, 98+%, Thermo Scientific Chemicals

Catalog number: L04258.06
5 g, Each
Thermo Scientific Chemicals

1,12-Diaminododecane, 98+%, Thermo Scientific Chemicals

Catalog number: L04258.06
5 g, Each
Quantity
Catalog number: L04258.06
also known as L04258-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2783-17-7
IUPAC Name
dodecane-1,12-diamine
Molecular Formula
C12H28N2
InChI Key
QFTYSVGGYOXFRQ-UHFFFAOYSA-N
SMILES
NCCCCCCCCCCCCN
Identification (FTIR)
Conforms
Appearance (Color)
White to cream to pale yellow
Assay (GC)
≥98.0%
Melting Point (clear melt)
67.0-73.0?C
Form
Crystals or powder or crystalline powder or chunks or fused solid or clear liquid as melt

Description

1,12-Diaminododecane is used for synthesis of medicinal drugs, polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,12-Diaminododecane is used for synthesis of medicinal drugs, polymers.

Solubility
Soluble in ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text