Isopropyl palmitate is used to formulate and evaluate the suitability of pluronic lecithin organogels containing flurbiprofen for topical application and to characterize microemulsion systems of isopropyl palmitate, water and 2:1 Brij 97, and 1-butanol by different experimental techniques.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Isopropyl palmitate is used to formulate and evaluate the suitability of pluronic lecithin organogels containing flurbiprofen for topical application and to characterize microemulsion systems of isopropyl palmitate, water and 2:1 Brij 97, and 1-butanol by different experimental techniques.
Solubility
Not miscible or difficult to mix with water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
- Prapaporn Boonme.; Karen Krauel.; Anja Graf.; Thomas Rades.; Varaporn Buraphacheep Junyaprasert. Characterization of microemulsion structures in the pseudoternary phase diagram of isopropyl palmitate/water/Brij 97:1-butanol. AAPS PharmSciTech. 2006, 7 (2), 99-104.
- H. Willimann.; P. Walde.; P. L. Luisi.; A. Gazzaniga.; F. Stroppolo. Lecithin organogel as matrix for transdermal transport of drugs. Journal of Pharmaceutical Sciences. 1992, 81 (9), 871-874.