Appearance (Color)White to pale grey to dark grey to pale brown to dark brown and/or grey-brown
Melting Point (clear melt)102-107°C
Identification (FTIR)Conforms
FormPowder or crystals or flakes
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.