Thermo Scientific Chemicals

Chlorocyclohexane, 98%, Thermo Scientific Chemicals

Catalog number: L04349.0B
1000 g, Each
Thermo Scientific Chemicals

Chlorocyclohexane, 98%, Thermo Scientific Chemicals

Catalog number: L04349.0B
1000 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: L04349.0B
also known as L04349-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
542-18-7
IUPAC Name
chlorocyclohexane
Molecular Formula
C6H11Cl
InChI Key
UNFUYWDGSFDHCW-UHFFFAOYSA-N
SMILES
ClC1CCCCC1
Appearance (Color)
Clear colorless to yellow
Identification (FTIR)
Conforms
Form
Liquid
Assay (GC)
≥97.5%
Refractive Index
1.4595-1.4645 @ 20?C

Description

Chlorocyclohexane is used as the raw material in synthesis agrochemical and medicine chemical engineering products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorocyclohexane is used as the raw material in synthesis agrochemical and medicine chemical engineering products.

Solubility
Soluble in water.0.02 g/L (20°C). Soluble in alcohol and diethyl ether.

Notes
Incompatible materials are Strong oxidizing agents, Strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text