1-Chlorooctane, 99%, Thermo Scientific Chemicals
1-Chlorooctane, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Chlorooctane, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number L04358.AE
also known as L04358-AE
Price (USD)/ Each
21.80
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
21.80
Add to cart
1-Chlorooctane, 99%, Thermo Scientific Chemicals
Catalog numberL04358.AE
Price (USD)/ Each
21.80
-
Add to cart
Chemical Identifiers
CAS111-85-3
IUPAC Name1-chlorooctane
Molecular FormulaC8H17Cl
InChI KeyCNDHHGUSRIZDSL-UHFFFAOYSA-N
SMILESCCCCCCCCCl
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥98.5%
Appearance (Color)Clear colorless
Refractive Index1.4285-1.4325 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only