Thermo Scientific Chemicals

1-Chlorooctane, 99%, Thermo Scientific Chemicals

Catalog number: L04358.AP
500 mL, Each
Thermo Scientific Chemicals

1-Chlorooctane, 99%, Thermo Scientific Chemicals

Catalog number: L04358.AP
500 mL, Each
Quantity
Catalog number: L04358.AP
also known as L04358-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
111-85-3
IUPAC Name
1-chlorooctane
Molecular Formula
C8H17Cl
InChI Key
CNDHHGUSRIZDSL-UHFFFAOYSA-N
SMILES
CCCCCCCCCl
Form
Liquid
Assay (GC)
≥98.5%
Appearance (Color)
Clear colorless
Refractive Index
1.4285-1.4325 @ 20?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text