Thermo Scientific Chemicals

Dichlorophenylsilane, 96%, Thermo Scientific Chemicals

Catalog number: L04371.06
5 g, Each
Thermo Scientific Chemicals

Dichlorophenylsilane, 96%, Thermo Scientific Chemicals

Catalog number: L04371.06
5 g, Each
Quantity
Catalog number: L04371.06
also known as L04371-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1631-84-1
IUPAC Name
dichloro(phenyl)silyl
Molecular Formula
C6H5Cl2Si
InChI Key
XNAFLNBULDHNJS-UHFFFAOYSA-N
SMILES
Cl[Si](Cl)C1=CC=CC=C1
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥95.0%

Description

Dichlorophenylsilane is used as the intermediates of OLED,pharmaceuticals, cosmetics. It is also used as reduction reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dichlorophenylsilane is used as the intermediates of OLED,pharmaceuticals, cosmetics. It is also used as reduction reagent.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Store away from moisture, oxidizing agents. Incompatible materials are bases, moisture and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text