N-Ethylmethylamine, 98+%, Thermo Scientific Chemicals
N-Ethylmethylamine, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Ethylmethylamine, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number L04372.14
also known as L04372-14
Price (USD)/ Each
193.65
Online exclusive
215.00 
Save 21.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
193.65
Online exclusive
215.00 
Save 21.35 (10%)
Add to cart
N-Ethylmethylamine, 98+%, Thermo Scientific Chemicals
Catalog numberL04372.14
Price (USD)/ Each
193.65
Online exclusive
215.00 
Save 21.35 (10%)
-
Add to cart
Chemical Identifiers
CAS624-78-2
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>98.0%
Refractive Index1.3720-1.3760 @ 20?C
Appearance (Color)Clear, colourless
Water Content (Karl Fischer Titration)<1%
Formliquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only