Thermo Scientific Chemicals

Methyl adipoyl chloride, 96%, Thermo Scientific Chemicals

Catalog number: L04377.09
10 g, Each
Thermo Scientific Chemicals

Methyl adipoyl chloride, 96%, Thermo Scientific Chemicals

Catalog number: L04377.09
10 g, Each
Quantity
Catalog number: L04377.09
also known as L04377-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
35444-44-1
IUPAC Name
methyl 6-chloro-6-oxohexanoate
Molecular Formula
C7H11ClO3
InChI Key
HDLGIEZOMYJKAK-UHFFFAOYSA-N
SMILES
COC(=O)CCCCC(Cl)=O
Assay (GC)
≥95.0%
Refractive Index
1.4445-1.4505 @ 20?C
Appearance (Color)
Clear colorless to white to yellow to orange to brown or pale pink
Assay (Titration ex Chloride)
≥95.0 to ≤105.0%
Identification (FTIR)
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text