n-Octyltriethoxysilane, 95%, Thermo Scientific Chemicals
n-Octyltriethoxysilane, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

n-Octyltriethoxysilane, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number L04407.18
also known as L04407-18
Price (USD)/ Each
20.90
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
20.90
Add to cart
n-Octyltriethoxysilane, 95%, Thermo Scientific Chemicals
Catalog numberL04407.18
Price (USD)/ Each
20.90
-
Add to cart
Chemical Identifiers
CAS2943-75-1
IUPAC Nametriethoxy(octyl)silane
Molecular FormulaC14H32O3Si
InChI KeyMSRJTTSHWYDFIU-UHFFFAOYSA-N
SMILESCCCCCCCC[Si](OCC)(OCC)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Refractive Index1.4135-1.4195 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥94.0%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only