Thermo Scientific Chemicals

n-Octyltriethoxysilane, 95%, Thermo Scientific Chemicals

Catalog number: L04407.30
250 g, Each
Thermo Scientific Chemicals

n-Octyltriethoxysilane, 95%, Thermo Scientific Chemicals

Catalog number: L04407.30
250 g, Each
Quantity
Catalog number: L04407.30
also known as L04407-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2943-75-1
IUPAC Name
triethoxy(octyl)silane
Molecular Formula
C14H32O3Si
InChI Key
MSRJTTSHWYDFIU-UHFFFAOYSA-N
SMILES
CCCCCCCC[Si](OCC)(OCC)OCC
Identification (FTIR)
Conforms
Refractive Index
1.4135-1.4195 @ 20?C
Appearance (Color)
Clear colorless
Assay (GC)
≥94.0%
Form
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text