Thermo Scientific Chemicals

4-Hydroxythiophenol, 97%, Thermo Scientific Chemicals

Catalog number: L04429.06
5 g, Each
Thermo Scientific Chemicals

4-Hydroxythiophenol, 97%, Thermo Scientific Chemicals

Catalog number: L04429.06
5 g, Each
Quantity
Catalog number: L04429.06
also known as L04429-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
637-89-8
IUPAC Name
4-sulfanylphenol
Molecular Formula
C6H6OS
InChI Key
BXAVKNRWVKUTLY-UHFFFAOYSA-N
SMILES
OC1=CC=C(S)C=C1
Appearance (Color)
White or colorless to cream to yellow
Assay (GC)
≥96.0%
Form
Crystalline or low melting/fused solid or clear liquid as melt

Description

4-Hydroxythiophenol is used to improve charge separation in PbSe quantum dots (QDs) as sensitizers for TiO2. It helps in the activation of neocarzinostatin-chromophore. Also used as a resin monomer in the study of the different gold nanocrystal core-resin shell structures

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxythiophenol is used to improve charge separation in PbSe quantum dots (QDs) as sensitizers for TiO2. It helps in the activation of neocarzinostatin-chromophore. Also used as a resin monomer in the study of the different gold nanocrystal core-resin shell structures

Solubility
Soluble in water.

Notes
Air Sensitive. Store at 4°C. Incompatible with air and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text