Thermo Scientific Chemicals

Triethanolamine, 98+%, Thermo Scientific Chemicals

Catalog number: L04486.36
500 g, Each
Thermo Scientific Chemicals

Triethanolamine, 98+%, Thermo Scientific Chemicals

Catalog number: L04486.36
500 g, Each
Quantity
Catalog number: L04486.36
Price (USD)
Have Questions?

Chemical Identifiers

CAS
102-71-6
IUPAC Name
2-[bis(2-hydroxyethyl)amino]ethan-1-ol
Molecular Formula
C6H15NO3
InChI Key
GSEJCLTVZPLZKY-UHFFFAOYSA-N
SMILES
OCCN(CCO)CCO
Assay (Non-aqueous acid-base Titration)
≥98.0 to ≤102.0%
Form
Viscous liquid
Identification (FTIR)
Conforms
Refractive Index
1.4830-1.4870 @ 20?C
Appearance (Color)
Clear colorless to pale yellow

Description

Protein purification reagent Triethanolamine acts as an organic additive in the grinding of cement clinker and as a complexing agent for aluminum ions in aqueous solutions. It is used as an emulsifier and a surfactant. It finds application in common products like dishwashing liquids, general cleaners, hand cleaners, shaving cream, metalworking fluids, liquid laundry detergents, paints and printing inks. It is used for the neutralization of fatty acids and adjusting buffer pH as well as utilized to dissolve oils. It acts as an effective complexing agent in electroless plating. Further, it is used in hair care products like hair dyes and wave sets.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Protein purification reagent Triethanolamine acts as an organic additive in the grinding of cement clinker and as a complexing agent for aluminum ions in aqueous solutions. It is used as an emulsifier and a surfactant. It finds application in common products like dishwashing liquids, general cleaners, hand cleaners, shaving cream, metalworking fluids, liquid laundry detergents, paints and printing inks. It is used for the neutralization of fatty acids and adjusting buffer pH as well as utilized to dissolve oils. It acts as an effective complexing agent in electroless plating. Further, it is used in hair care products like hair dyes and wave sets.

Solubility
Miscible with water, methanol, acetone, ether, carbon tetrachloride, n-heptane and chloroform. Slightly soluble in petroleum ether.

Notes
Air sensitive and hygroscopic Incompatible with acids, oxidizing agents, reducing agents and metals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text