2-Picoline N-oxide is used in proteomics research. Also used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Picoline N-oxide is used in proteomics research. Also used as pharmaceutical intermediates.
Solubility
Soluble in water.
Notes
Hygroscopic. Store under dry inert gas. Incompatible with acids, Acid chlorides, Acid anhydrides, Chloroformates.
RUO – Research Use Only
Shigeru. Oae; Teijiro. Kitao; Yoshinori. Kitaoka. The Mechanism of the Reaction of 2-Picoline N-Oxide with Acetic Anhydride.J. Am. Chem. Soc.1962, 84 (17), 3359-3362.
R. S. Sagerr; W. H. Watson. 2-Picoline N-oxide complexes of copper(II)chloride. The crystal structure of Cu3 Cl6(C6H7NO)2.2H2O.Inorg. Chem.1968, 7 (10), 2035-2040.