Thermo Scientific Chemicals

1,1,2,2-Tetrachloroethane, 98+%, Thermo Scientific Chemicals

Catalog number: L04489.22
100 g, Each
Thermo Scientific Chemicals

1,1,2,2-Tetrachloroethane, 98+%, Thermo Scientific Chemicals

Catalog number: L04489.22
100 g, Each
Quantity
Catalog number: L04489.22
also known as L04489-22
Price (USD)
Have Questions?

Chemical Identifiers

CAS
79-34-5
IUPAC Name
1,1,2,2-tetrachloroethane
Molecular Formula
C2H2Cl4
InChI Key
QPFMBZIOSGYJDE-UHFFFAOYSA-N
SMILES
ClC(Cl)C(Cl)Cl
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay from Suppliers CofA
≥98.0%

Description

1,1,2,2-Tetrachloroethane is used in the preparation of organic chemicals, as a solvent for fats, oils, waxes, resins, cellulose acetate and in certain types of Friedel-Crafts reactions. It is also used to make paints, varnishes, rust removers and other chlorinated hydrocarbons with two carbon atoms. It is utilized in the determination of theobromine in cacao. It is also used as an immersion fluid in crystallography. Further, it is used as an alcohol denaturant, fumigant, dry cleaning agent, in photographic film. In addition to this, it is used as an intermediate in the industrial production of trichloroethylene, tetrachloroethylene and1,2-dichloroethylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1,2,2-Tetrachloroethane is used in the preparation of organic chemicals, as a solvent for fats, oils, waxes, resins, cellulose acetate and in certain types of Friedel-Crafts reactions. It is also used to make paints, varnishes, rust removers and other chlorinated hydrocarbons with two carbon atoms. It is utilized in the determination of theobromine in cacao. It is also used as an immersion fluid in crystallography. Further, it is used as an alcohol denaturant, fumigant, dry cleaning agent, in photographic film. In addition to this, it is used as an intermediate in the industrial production of trichloroethylene, tetrachloroethylene and1,2-dichloroethylene.

Solubility
Miscible with water, methanol, ethanol, chloroform and acetone.

Notes
Incompatible with strong oxidizing agents, sodium, sodium oxides, strong bases, potassium, nitrates, 2,4-dinitrophenyl disulfide.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text