Thermo Scientific Chemicals

1,2,4,5-Tetramethylbenzene, 97+%, Thermo Scientific Chemicals

Catalog number: L04493.36
500 g, Each
Thermo Scientific Chemicals

1,2,4,5-Tetramethylbenzene, 97+%, Thermo Scientific Chemicals

Catalog number: L04493.36
500 g, Each
Quantity
Catalog number: L04493.36
also known as L04493-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
95-93-2
IUPAC Name
1,2,4,5-tetramethylbenzene
Molecular Formula
C10H14
InChI Key
SQNZJJAZBFDUTD-UHFFFAOYSA-N
SMILES
CC1=CC(C)=C(C)C=C1C
Appearance (Color)
White
Identification (FTIR)
Conforms
Melting Point (clear melt)
76-82?C
Form
Chunks or powder
Assay (GC)
≥97.0%

Description

1,2,4,5-Tetramethylbenzene is the precursor to pyromellitic dianhydride, which is used for manufacturing curing agents, adhesives, coating materials. It is used in the manufacture of some raw materials for engineering plastics (polyimides) and cross-linking agent for alkyd resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,4,5-Tetramethylbenzene is the precursor to pyromellitic dianhydride, which is used for manufacturing curing agents, adhesives, coating materials. It is used in the manufacture of some raw materials for engineering plastics (polyimides) and cross-linking agent for alkyd resins.

Solubility
Soluble in alcohol, ether, benzene, and acetone. Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text