Thermo Scientific Chemicals

N,N-Diethylformamide, 99%, Thermo Scientific Chemicals

Catalog number: L04503.22
100 g, Each
Thermo Scientific Chemicals

N,N-Diethylformamide, 99%, Thermo Scientific Chemicals

Catalog number: L04503.22
100 g, Each
Quantity
Catalog number: L04503.22
also known as L04503-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
617-84-5
IUPAC Name
N,N-diethylformamide
Molecular Formula
C5H11NO
InChI Key
SUAKHGWARZSWIH-UHFFFAOYSA-N
SMILES
CCN(CC)C=O
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Refractive Index
1.4325-1.4355 @ 20?C

Description

N,N-Diethylformamide is used in the synthesis of metal-organic frameworks and Diethyltrifluoromethylamine. It is also used as solvent in the preparation of porous cubic-shaped zinc oxide particles. It is involved in the preparation of quinazoline-2,4(1H,3H)-dione by reacting with o-aminonitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Diethylformamide is used in the synthesis of metal-organic frameworks and Diethyltrifluoromethylamine. It is also used as solvent in the preparation of porous cubic-shaped zinc oxide particles. It is involved in the preparation of quinazoline-2,4(1H,3H)-dione by reacting with o-aminonitrile.

Solubility
Miscible with acetone, benzene, alcohol and ether.  

Notes
Incompatible with strong oxidizing agents. Avoid heat, flames and spark.                      
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text