Thermo Scientific Chemicals

Dimethylphenylsilane, 97%, Thermo Scientific Chemicals

Catalog number: L04558.09
10 g, Each
Thermo Scientific Chemicals

Dimethylphenylsilane, 97%, Thermo Scientific Chemicals

Catalog number: L04558.09
10 g, Each
Quantity
Catalog number: L04558.09
also known as L04558-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
766-77-8
IUPAC Name
dimethyl(phenyl)silyl
Molecular Formula
C8H11Si
InChI Key
OIKHZBFJHONJJB-UHFFFAOYSA-N
SMILES
C[Si](C)C1=CC=CC=C1
Form
Liquid
Refractive Index
1.4955-1.5005 @ 20?C
Assay (GC)
≥96.0%
Appearance (Color)
Clear colorless

Description

Dimethylphenylsilane is a reagent for enol ether synthesis. It acts as a catalyst. Also used as a precursor in Optical Emission Spectroscopy of Plasma Deposition Processes. It can react with vinylbenzene to produce (b-Phenyl-ethyl)-dimethylphenyl-silan.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethylphenylsilane is a reagent for enol ether synthesis. It acts as a catalyst. Also used as a precursor in Optical Emission Spectroscopy of Plasma Deposition Processes. It can react with vinylbenzene to produce (b-Phenyl-ethyl)-dimethylphenyl-silan.

Solubility
Not miscible in water.

Notes
Moisture sensitive. Incompatible materials are Strong oxidizing agents, Strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text