1-Phenoxy-2-propanol, tech. 85%, Thermo Scientific Chemicals
1-Phenoxy-2-propanol, tech. 85%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Phenoxy-2-propanol, tech. 85%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number L04596.30
also known as L04596-30
Price (USD)/ Each
115.65
Online exclusive
128.00 
Save 12.35 (10%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
115.65
Online exclusive
128.00 
Save 12.35 (10%)
Add to cart
1-Phenoxy-2-propanol, tech. 85%, Thermo Scientific Chemicals
Catalog numberL04596.30
Price (USD)/ Each
115.65
Online exclusive
128.00 
Save 12.35 (10%)
-
Add to cart
Chemical Identifiers
CAS770-35-4
IUPAC Name1-phenoxypropan-2-ol
Molecular FormulaC9H12O2
InChI KeyIBLKWZIFZMJLFL-UHFFFAOYNA-N
SMILESCC(O)COC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colourless
Assay (GC)>83.0%
Formliquid
Impurity content2-Phenoxy-1-propanol: <15.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only