Thermo Scientific Chemicals

1,5-Dihydroxynaphthalene, 98%, Thermo Scientific Chemicals

Catalog number: L04599.30
250 g, Each
Thermo Scientific Chemicals

1,5-Dihydroxynaphthalene, 98%, Thermo Scientific Chemicals

Catalog number: L04599.30
250 g, Each
Quantity
Catalog number: L04599.30
also known as L04599-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
83-56-7
IUPAC Name
naphthalene-1,5-diol
Molecular Formula
C10H8O2
InChI Key
BOKGTLAJQHTOKE-UHFFFAOYSA-N
SMILES
OC1=CC=CC2=C(O)C=CC=C12
Appearance (Color)
Dark brown to brown to gray
Assay (HPLC)
≥97.5%
Form
Powder
Identification (FTIR)
Conforms

Description

1,5-Dihydroxynaphthalene is an intermediate of synthetic mordant azo dyes. It is an intermediate used in organic synthesis, pharmaceuticals, dyestuff fields and photograph industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,5-Dihydroxynaphthalene is an intermediate of synthetic mordant azo dyes. It is an intermediate used in organic synthesis, pharmaceuticals, dyestuff fields and photograph industry.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text