Thermo Scientific Chemicals

1-Bromo-4-cyclohexylbenzene, 98%, Thermo Scientific Chemicals

Catalog number: L04636.06
5 g, Each
Thermo Scientific Chemicals

1-Bromo-4-cyclohexylbenzene, 98%, Thermo Scientific Chemicals

Catalog number: L04636.06
5 g, Each
Quantity
Catalog number: L04636.06
also known as L04636-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
25109-28-8
IUPAC Name
1-bromo-4-cyclohexylbenzene
Molecular Formula
C12H15Br
InChI Key
LVIJLEREXMVRAN-UHFFFAOYSA-N
SMILES
BrC1=CC=C(C=C1)C1CCCCC1
Form
Liquid
Appearance (Color)
Clear colorless
Refractive Index
1.5580-1.5620 @ 20?C
Assay (GC)
≥97.5%

Description

1-Bromo-4-cyclohexylbenzene is used as a organic light-emitting diode and as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-4-cyclohexylbenzene is used as a organic light-emitting diode and as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text