Thermo Scientific Chemicals

2,3-Hexanedione, 94%, Thermo Scientific Chemicals

Catalog number: L04669.06
5 g, Each
Thermo Scientific Chemicals

2,3-Hexanedione, 94%, Thermo Scientific Chemicals

Catalog number: L04669.06
5 g, Each
Quantity
Catalog number: L04669.06
also known as L04669-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3848-24-6
IUPAC Name
hexane-2,3-dione
Molecular Formula
C6H10O2
InChI Key
MWVFCEVNXHTDNF-UHFFFAOYSA-N
SMILES
CCCC(=O)C(C)=O
Identification (FTIR)
Conforms
Appearance (Color)
Clear pale yellow to yellow
Assay (GC)
≥92.0%
Refractive Index
1.4075-1.4165 @ 20?C
Form
Liquid

Description

2,3-Hexanedione is used to evaluate the influence of xanthan concentration on the release of aroma compounds in xanthan-thickened food model systems. It reacts with ethylenediamine to yield macrocyclic tetradentate 12-membered nitrogen donor (N4) ligand.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Hexanedione is used to evaluate the influence of xanthan concentration on the release of aroma compounds in xanthan-thickened food model systems. It reacts with ethylenediamine to yield macrocyclic tetradentate 12-membered nitrogen donor (N4) ligand.

Solubility
Partly miscible in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text