Thermo Scientific Chemicals

4-Chlorotoluene, 98%, Thermo Scientific Chemicals

Catalog number: L04672.36
500 g, Each
Thermo Scientific Chemicals

4-Chlorotoluene, 98%, Thermo Scientific Chemicals

Catalog number: L04672.36
500 g, Each
Quantity
Catalog number: L04672.36
also known as L04672-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
106-43-4
IUPAC Name
1-chloro-4-methylbenzene
Molecular Formula
C7H7Cl
InChI Key
NPDACUSDTOMAMK-UHFFFAOYSA-N
SMILES
CC1=CC=C(Cl)C=C1
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥97.5%
Refractive Index
1.5190-1.5230 @ 20?C
Form
Liquid

Description

4-Chlorotoluene is used for the preparation model gas of 4-chlorotoulene by passing synthesized air into liquid 4-chlorotoulene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorotoluene is used for the preparation model gas of 4-chlorotoulene by passing synthesized air into liquid 4-chlorotoulene.

Solubility
Soluble in water (0.04 g/L at 20oc).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text