Thermo Scientific Chemicals

Phenyltriethoxysilane, 98%, Thermo Scientific Chemicals

Catalog number: L04684.18
50 g, Each
Thermo Scientific Chemicals

Phenyltriethoxysilane, 98%, Thermo Scientific Chemicals

Catalog number: L04684.18
50 g, Each
Quantity
Catalog number: L04684.18
also known as L04684-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
780-69-8
IUPAC Name
triethoxy(phenyl)silane
Molecular Formula
C12H20O3Si
InChI Key
JCVQKRGIASEUKR-UHFFFAOYSA-N
SMILES
CCO[Si](OCC)(OCC)C1=CC=CC=C1
Assay (GC)
≥97.5%
Appearance (Color)
Clear colorless
Refractive Index
1.4590-1.4630 @ 20?C
Form
Liquid or viscous liquid
Identification (FTIR)
Conforms

Description

Phenyltriethoxysilane is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyltriethoxysilane is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Hydrolyzes in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture. Store under dry inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text