Thermo Scientific Chemicals

4-Nitro-o-phenylenediamine, 97%, Thermo Scientific Chemicals

Catalog number: L04693.22
100 g, Each
Thermo Scientific Chemicals

4-Nitro-o-phenylenediamine, 97%, Thermo Scientific Chemicals

Catalog number: L04693.22
100 g, Each
Quantity
Catalog number: L04693.22
also known as L04693-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
99-56-9
IUPAC Name
4-nitrobenzene-1,2-diamine
Molecular Formula
C6H7N3O2
InChI Key
RAUWPNXIALNKQM-UHFFFAOYSA-N
SMILES
NC1=CC=C(C=C1N)[N+]([O-])=O
Appearance (Color)
Orange to red to dark red or red-brown to brown
Identification (FTIR)
Conforms
Assay (Non-aqueous acid-base Titration)
≥96.0 to ≤104.0%
Melting Point (clear melt)
199-207?C
Form
Powder

Description

4-Nitro-o-phenylenediamine is used to study the in vitro activation of 4-nitro-o-phenylenediamine into a potent mutagen in Salmonella typhimurium strain TA98

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitro-o-phenylenediamine is used to study the in vitro activation of 4-nitro-o-phenylenediamine into a potent mutagen in Salmonella typhimurium strain TA98

Solubility
Slightly soluble in water (1.2 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text