Thermo Scientific Chemicals

1-(3-Aminopropyl)pyrrolidine, 97%, Thermo Scientific Chemicals

Catalog number: L04739.14
25 g, Each
Thermo Scientific Chemicals

1-(3-Aminopropyl)pyrrolidine, 97%, Thermo Scientific Chemicals

Catalog number: L04739.14
25 g, Each
Quantity
Catalog number: L04739.14
also known as L04739-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
23159-07-1
IUPAC Name
3-(pyrrolidin-1-yl)propan-1-amine
Molecular Formula
C7H16N2
InChI Key
VPBWZBGZWHDNKL-UHFFFAOYSA-N
SMILES
NCCCN1CCCC1
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥96.0%
Refractive Index
1.4685-1.4735 @ 20?C

Description

1-(3-Aminopropyl)pyrrolidine is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It can be used to make other chemicals It can react with 7-chloro-6-(2-chloro-ethyl)-5-methyl-[1,2,4]triazolo[1,5-a]pyrimidine to get 5-methyl-8-(3-pyrrolidin-1-yl-propyl)-7,8-dihydro-6H-1,3,4,8,8b-pentaaza-as-indacene . It can also react with 2,4-diphenyl-cyclobutane-1,3-dicarbonyl dichloride to obtain 2,4-diphenyl-cyclobutane-1,3-dicarboxylic acid bis-[(3-pyrrolidin-1-yl-propyl)-amide] .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(3-Aminopropyl)pyrrolidine is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It can be used to make other chemicals It can react with 7-chloro-6-(2-chloro-ethyl)-5-methyl-[1,2,4]triazolo[1,5-a]pyrimidine to get 5-methyl-8-(3-pyrrolidin-1-yl-propyl)-7,8-dihydro-6H-1,3,4,8,8b-pentaaza-as-indacene . It can also react with 2,4-diphenyl-cyclobutane-1,3-dicarbonyl dichloride to obtain 2,4-diphenyl-cyclobutane-1,3-dicarboxylic acid bis-[(3-pyrrolidin-1-yl-propyl)-amide] .

Solubility
Soluble in water, soluble in chloroform

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text