Thermo Scientific Chemicals

N,N-Diisopropylethylenediamine, 97%, Thermo Scientific Chemicals

Catalog number: L04765.18
50 g, Each
Thermo Scientific Chemicals

N,N-Diisopropylethylenediamine, 97%, Thermo Scientific Chemicals

Catalog number: L04765.18
50 g, Each
Quantity
Catalog number: L04765.18
also known as L04765-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
121-05-1
IUPAC Name
(2-aminoethyl)bis(propan-2-yl)amine
Molecular Formula
C8H20N2
InChI Key
CURJNMSGPBXOGK-UHFFFAOYSA-N
SMILES
CC(C)N(CCN)C(C)C
Appearance (Color)
Clear, colourless to yellow
Form
liquid
Assay (GC)
>96.0%
Refractive Index
1.4395-1.4445

Description

N,N-Diisopropylethylenediamine can be used to produce 1,4-diisopropyl-piperazin-2-one by heating. It is used as ligands for metathesis catalysts. It acts as anti-microbial agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Diisopropylethylenediamine can be used to produce 1,4-diisopropyl-piperazin-2-one by heating. It is used as ligands for metathesis catalysts. It acts as anti-microbial agent.

Solubility
Fully miscible in water.

Notes
Air Sensitive. Store away from air, oxidizing agents. Incompatible materials are Strong oxidizing agents, Strong acids
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text