Thermo Scientific Chemicals

Cholesteryl chloroformate, Thermo Scientific Chemicals

Catalog number: L04801.14
25 g, Each
Thermo Scientific Chemicals

Cholesteryl chloroformate, Thermo Scientific Chemicals

Catalog number: L04801.14
25 g, Each
Quantity
Catalog number: L04801.14
also known as L04801-14
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7144-08-3
IUPAC Name
(1R,3aS,3bS,7S,9aR,9bS,11aR)-9a,11a-dimethyl-1-[(2R)-6-methylheptan-2-yl]-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-yl carbonochloridate
Molecular Formula
C28H45ClO2
InChI Key
QNEPTKZEXBPDLF-JDTILAPWSA-N
SMILES
CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@H](CC[C@]4(C)[C@H]3CC[C@]12C)OC(Cl)=O
Appearance (Color)
White to pale cream
Purity (DSC)
≥97.5% (non-U.S. sourced material)
Melting Point
118.0-120.5°C (U.S. sourced material)
Infrared spectrum
Conforms (U.S. Sourced material)
Optical Rotation
-29° ± 2° (c=5 in Chloroform, non-U.S. sourced material)

Description

Cholesteryl chloroformate is used in the preparation of hydrophobized chitosan oligosaccharide, which finds application as an efficient gene carrier. It acts as an initiator in the polymerization of methyl methacrylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cholesteryl chloroformate is used in the preparation of hydrophobized chitosan oligosaccharide, which finds application as an efficient gene carrier. It acts as an initiator in the polymerization of methyl methacrylate.

Solubility
Soluble in methylene chloride, chloroform, acetone, toluene and benzene. Slightly soluble in methanol and ethanol.

Notes
Moisture sensitive. Incompatible with strong bases and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text