Thermo Scientific Chemicals

1-(3-Aminopropyl)-4-methylpiperazine, 98%, Thermo Scientific Chemicals

Catalog number: L04876.14
25 g, Each
Thermo Scientific Chemicals

1-(3-Aminopropyl)-4-methylpiperazine, 98%, Thermo Scientific Chemicals

Catalog number: L04876.14
25 g, Each
Quantity
Catalog number: L04876.14
also known as L04876-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4572-03-6
IUPAC Name
3-(4-methylpiperazin-1-yl)propan-1-amine
Molecular Formula
C8H19N3
InChI Key
RGUABPVONIGVAT-UHFFFAOYSA-N
SMILES
CN1CCN(CCCN)CC1
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
>97.5%
Refractive Index
1.4800-1.4840 @ 20?C

Description

1-(3-Aminopropyl)-4-methylpiperazine is used as a precursor in organic synthesis for the preparation of furan-2-carboxylic acid [3-(4-methyl-piperazin-1-yl)-propyl]-amide by reacting with furan-2-carbonyl chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(3-Aminopropyl)-4-methylpiperazine is used as a precursor in organic synthesis for the preparation of furan-2-carboxylic acid [3-(4-methyl-piperazin-1-yl)-propyl]-amide by reacting with furan-2-carbonyl chloride.

Notes
Air sensitive. Keep in a dry, cool and well-ventilated place. Keep container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text