Indole-3-propionic acid, 98%
Indole-3-propionic acid, 98%
Indole-3-propionic acid, 98%
Indole-3-propionic acid, 98%
Thermo Scientific Chemicals

Indole-3-propionic acid, 98%

CAS: 830-96-6 | C11H11NO2 | 189.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number L04877.14
also known as L04877-14
Price (USD)
121.00
Each
Estimated availability date 10-Jul-2025
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
121.00
Each
Add to cart
Chemical Identifiers
CAS830-96-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream or pale yellow or pale brown
FormPowder
Assay from Supplier's CofA≥97.5%
Identification (FTIR)Conforms
Indole-3-propionic acid is a reactant for preparation of fluorescent analogues of strigolactones, melanocortin receptors ligands, histamine H4 receptor agonists, and NR2B/NMDA receptor antagonists. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used to Study as an adjunct to improve perfusion after liver transplant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Indole-3-propionic acid is a reactant for preparation of fluorescent analogues of strigolactones, melanocortin receptors ligands, histamine H4 receptor agonists, and NR2B/NMDA receptor antagonists. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used to Study as an adjunct to improve perfusion after liver transplant.

Solubility
It is slightly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Yuan-Hsiao Su et. al. Solution-phase parallel synthesis and screening of anti-tumor activities from fenbufen and ethacrynic acid libraries. Bioorganic & Medicinal Chemistry Letters.2011, 21,(5), 1320-1324 .
  2. Francis Giraud et. al. Synthesis ofN-aryl-3-(indol-3-yl)propanamides and their immunosuppressive activities. Bioorganic & Medicinal Chemistry Letters.2011, 20,(17), 5203-5206.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link