Thermo Scientific Chemicals

2-Mercaptothiazoline, 98+%, Thermo Scientific Chemicals

Catalog number: L04924.22
100 g, Each
Thermo Scientific Chemicals

2-Mercaptothiazoline, 98+%, Thermo Scientific Chemicals

Catalog number: L04924.22
100 g, Each
Quantity
Catalog number: L04924.22
also known as L04924-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
96-53-7
IUPAC Name
1,3-thiazolidine-2-thione
Molecular Formula
C3H5NS2
InChI Key
WGJCBBASTRWVJL-UHFFFAOYSA-N
SMILES
S=C1NCCS1
Appearance (Color)
White to cream or pale yellow
Assay (GC)
≥98.0%
Form
Powder or crystalline powder
Identification (FTIR)
Conforms
Melting Point (clear melt)
100-108?C

Description

2-Mercaptothiazoline, is used in the chain-lengthening of alkyl halides by trans-iodopropenylenation. 2-Mercaptothiazoline is an antithyroid agent that strongly reduces thyroid hormone levels. 2-Mercaptothiazoline is also used as an intermediate in the preparation of biologically active compounds. It is also used as a tool for highly selective chiral synthesis of penam- and carbapenam-type β-lactam antibiotics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Mercaptothiazoline, is used in the chain-lengthening of alkyl halides by trans-iodopropenylenation. 2-Mercaptothiazoline is an antithyroid agent that strongly reduces thyroid hormone levels. 2-Mercaptothiazoline is also used as an intermediate in the preparation of biologically active compounds. It is also used as a tool for highly selective chiral synthesis of penam- and carbapenam-type β-lactam antibiotics.

Solubility
Soluble in chloroform, methanol. Slightly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text