Thermo Scientific Chemicals

(+)-alpha-Pinene, 98%, Thermo Scientific Chemicals

Catalog number: L04941.30
250 g, Each
Thermo Scientific Chemicals

(+)-alpha-Pinene, 98%, Thermo Scientific Chemicals

Catalog number: L04941.30
250 g, Each
Quantity
Catalog number: L04941.30
also known as L04941-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7785-70-8
IUPAC Name
(1R,5R)-2,6,6-trimethylbicyclo[3.1.1]hept-2-ene
Molecular Formula
C10H16
InChI Key
GRWFGVWFFZKLTI-RKDXNWHRSA-N
SMILES
CC1=CC[C@@H]2C[C@H]1C2(C)C
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
≥97.5%
Optical Rotation
40.0? to 48.0? (neat)
Refractive Index
1.4635-1.4680 @ 20?C

Description

(+)-alpha-Pinene is employed in the preparation of chiral hydroboration reagents. It is a bronchodilator in humans. α-Pinene is an anti-inflammatory and used as a broad-spectrum antibiotic.It exhibits activity as an acetylcholinesterase inhibitor, aiding memory. It is a biosynthetic base for CB2 ligands, such as HU-308.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+)-alpha-Pinene is employed in the preparation of chiral hydroboration reagents. It is a bronchodilator in humans. α-Pinene is an anti-inflammatory and used as a broad-spectrum antibiotic.It exhibits activity as an acetylcholinesterase inhibitor, aiding memory. It is a biosynthetic base for CB2 ligands, such as HU-308.

Solubility
Soluble in Ether, Alcohols, Chloroform. Not miscible in water.

Notes
Incompatible materials are Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text