Thermo Scientific Chemicals

meso-1,2-Dibromo-1,2-diphenylethane, 97%, Thermo Scientific Chemicals

Catalog number: L04956.06
5 g, Each
Thermo Scientific Chemicals

meso-1,2-Dibromo-1,2-diphenylethane, 97%, Thermo Scientific Chemicals

Catalog number: L04956.06
5 g, Each
Quantity
Catalog number: L04956.06
also known as L04956-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13440-24-9
IUPAC Name
(1,2-dibromo-2-phenylethyl)benzene
Molecular Formula
C14H12Br2
InChI Key
GKESIQQTGWVOLH-UHFFFAOYNA-N
SMILES
BrC(C(Br)C1=CC=CC=C1)C1=CC=CC=C1
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥96.0%
Identification (FTIR)
Conforms
Appearance (Color)
White to pale cream

Description

meso-1,2-Dibromo-1,2-diphenylethane is used to study the reaction of ± and meso-SBr2 with 9-substituted fluorenide ions in dimethyl sulfoxide. It undergoes electrochemical dehalogenation in acetonitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
meso-1,2-Dibromo-1,2-diphenylethane is used to study the reaction of ± and meso-SBr2 with 9-substituted fluorenide ions in dimethyl sulfoxide. It undergoes electrochemical dehalogenation in acetonitrile.

Solubility
Insoluble in water.

Notes
Incompatible materials are Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text