7-Phenylheptanoic acid is used in the process to make amino acid derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
7-Phenylheptanoic acid is used in the process to make amino acid derivatives.
Solubility
Insoluble in water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
RUO – Research Use Only
TM Harris.; CM Harris. Lactonization of 3, 5, 7-trioxo-7-phenylheptanoic acid and its 2-methyl homolog. Tetrahedron. 1969, 25,(13), 2687-2691.
B Hazer.; RW Lenz.; RC Fuller. Bacterial production of poly-3-hydroxyalkanoates containing arylalkyl substituent groups. Polymer. 1996, 37,(26), 5951-5957.