Thermo Scientific Chemicals

Succinonitrile, 98%, Thermo Scientific Chemicals

Catalog number: L05037.22
100 g, Each
Thermo Scientific Chemicals

Succinonitrile, 98%, Thermo Scientific Chemicals

Catalog number: L05037.22
100 g, Each
Quantity
Catalog number: L05037.22
also known as L05037-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
110-61-2
IUPAC Name
butanedinitrile
Molecular Formula
C4H4N2
InChI Key
IAHFWCOBPZCAEA-UHFFFAOYSA-N
SMILES
N#CCCC#N
Appearance (Color)
White to pale cream to cream to brown to gray
Form
Crystals or powder or crystalline powder or lumps or fused solid or waxy solid
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms

Description

Succinonitrile dispersed with ionic liquids entrapped in a host polymer was used to constitute the gel polymer electrolytes. It was used as dopant to investigate the ionic conductivity and X-ray absorption spectroscopic results for lithium and copper salt doped succinonitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Succinonitrile dispersed with ionic liquids entrapped in a host polymer was used to constitute the gel polymer electrolytes. It was used as dopant to investigate the ionic conductivity and X-ray absorption spectroscopic results for lithium and copper salt doped succinonitrile.

Solubility
Slightly soluble in methanol and water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text