Succinonitrile, 98%, Thermo Scientific Chemicals
Succinonitrile, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Succinonitrile, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number L05037.36
also known as L05037-36
Price (USD)/ Each
161.00
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
161.00
Add to cart
Succinonitrile, 98%, Thermo Scientific Chemicals
Catalog numberL05037.36
Price (USD)/ Each
161.00
-
Add to cart
Chemical Identifiers
CAS110-61-2
IUPAC Namebutanedinitrile
Molecular FormulaC4H4N2
InChI KeyIAHFWCOBPZCAEA-UHFFFAOYSA-N
SMILESN#CCCC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream to cream to brown to gray
FormCrystals or powder or crystalline powder or lumps or fused solid or waxy solid
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Succinonitrile dispersed with ionic liquids entrapped in a host polymer was used to constitute the gel polymer electrolytes. It was used as dopant to investigate the ionic conductivity and X-ray absorption spectroscopic results for lithium and copper salt doped succinonitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Succinonitrile dispersed with ionic liquids entrapped in a host polymer was used to constitute the gel polymer electrolytes. It was used as dopant to investigate the ionic conductivity and X-ray absorption spectroscopic results for lithium and copper salt doped succinonitrile.

Solubility
Slightly soluble in methanol and water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, strong acids.
RUO – Research Use Only

General References:

  1. Mohd Suleman et. al. Structural and electrochemical properties of succinonitrile-based gel polymer electrolytes: role of ionic liquid addition. Journal of Physical Chemistry B. 2013, 117,(24), 7436-7443 .
  2. Michaela K Jahn et. al. Substituent steering of dihedral angles around single bonds: the case of succinonitrile. Physical Chemistry Chemical Physics. 2014, 16,(5), 2100-2105.