N,N-Dimethylurea, 98%, Thermo Scientific Chemicals
N,N-Dimethylurea, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,N-Dimethylurea, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number L05065.18
also known as L05065-18
Price (USD)/ Each
135.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
135.00
Add to cart
N,N-Dimethylurea, 98%, Thermo Scientific Chemicals
Catalog numberL05065.18
Price (USD)/ Each
135.00
-
Add to cart
Chemical Identifiers
CAS598-94-7
IUPAC Name1,1-dimethylurea
Molecular FormulaC3H8N2O
InChI KeyYBBLOADPFWKNGS-UHFFFAOYSA-N
SMILESCN(C)C(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥97.5%
Appearance (Color)White
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only