Thermo Scientific Chemicals

7-Bromoheptanenitrile, 98%, Thermo Scientific Chemicals

Catalog number: L05068.14
25 g, Each
Thermo Scientific Chemicals

7-Bromoheptanenitrile, 98%, Thermo Scientific Chemicals

Catalog number: L05068.14
25 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: L05068.14
also known as L05068-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
20965-27-9
IUPAC Name
7-bromoheptanenitrile
Molecular Formula
C7H12BrN
InChI Key
HVVQSKCGHAPHMV-UHFFFAOYSA-N
SMILES
BrCCCCCCC#N
Appearance (Color)
Clear colorless to white to yellow to pale orange
Form
Liquid
Assay (GC)
≥97.5%
Refractive Index
1.4730-1.4770 @ 20?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text