Thermo Scientific Chemicals

1-Bromo-2-methylnaphthalene, tech. 90%, Thermo Scientific Chemicals

Catalog number: L05093.09
10 g, Each
Thermo Scientific Chemicals

1-Bromo-2-methylnaphthalene, tech. 90%, Thermo Scientific Chemicals

Catalog number: L05093.09
10 g, Each
Quantity
Catalog number: L05093.09
also known as L05093-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2586-62-1
IUPAC Name
1-bromo-2-methylnaphthalene
Molecular Formula
C11H9Br
InChI Key
CMIMBQIBIZZZHQ-UHFFFAOYSA-N
SMILES
CC1=CC=C2C=CC=CC2=C1Br
Form
liquid
Assay (GC)
> 88.0%
Refractive Index
1.6450 - 1.6530 @20?C
Appearance (Color)
Clear colourless or yellow to orange

Description

1-Bromo-2-methylnaphthalene is used as test compound in determination of halogenated hydrocarbons at trace levels by supercritical fluid chromatography-microwave-induced plasma mass spectrometry. Dynamic phosphorescence quenching of 1-bromo-2-methylnaphthalene without deoxygenation has been used for selective sensing of Cu(II) at ngm levels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2-methylnaphthalene is used as test compound in determination of halogenated hydrocarbons at trace levels by supercritical fluid chromatography-microwave-induced plasma mass spectrometry. Dynamic phosphorescence quenching of 1-bromo-2-methylnaphthalene without deoxygenation has been used for selective sensing of Cu(II) at ngm levels.

Solubility
Not miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text