Thermo Scientific Chemicals

2-Naphthoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: L05102.14
25 g, Each
Thermo Scientific Chemicals

2-Naphthoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: L05102.14
25 g, Each
Quantity
Catalog number: L05102.14
also known as L05102-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
93-09-4
IUPAC Name
naphthalene-2-carboxylic acid
Molecular Formula
C11H8O2
InChI Key
UOBYKYZJUGYBDK-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=C2C=CC=CC2=C1
Appearance (Color)
White to pale cream to pale brown
Form
Powder
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Assay (Silylated GC)
≥98.0%
Identification (FTIR)
Conforms

Description

2-Naphthoic acid is an unprotected acid undergoes lithiation, e.g. with s-BuLi, by stereospecific 1,4-addition, providing a facile route to 1,2,2-trisubstituted 1,2-dihydronaphthalenes. It is also used as an intermediate in organic synthesis. The fluorescence spectra and electronic absorption of 2-naphthoic acid was studied.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Naphthoic acid is an unprotected acid undergoes lithiation, e.g. with s-BuLi, by stereospecific 1,4-addition, providing a facile route to 1,2,2-trisubstituted 1,2-dihydronaphthalenes. It is also used as an intermediate in organic synthesis. The fluorescence spectra and electronic absorption of 2-naphthoic acid was studied.

Solubility
Soluble in alcohol, diethyl ether, methanol, and ethyl acetate (20 mg/ml). Insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text