Thermo Scientific Chemicals

Methyl cholate, 97%, Thermo Scientific Chemicals

Catalog number: L05129.06
5 g, Each
Thermo Scientific Chemicals

Methyl cholate, 97%, Thermo Scientific Chemicals

Catalog number: L05129.06
5 g, Each
Quantity
Catalog number: L05129.06
also known as L05129-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1448-36-8
IUPAC Name
methyl 4-{4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl}pentanoate
Molecular Formula
C25H42O5
InChI Key
DLYVTEULDNMQAR-UHFFFAOYNA-N
SMILES
COC(=O)CCC(C)C1CCC2C3C(O)CC4CC(O)CCC4(C)C3CC(O)C12C
Appearance (Color)
White to cream
Form
Crystals or powder or crystalline powder
Assay (special)
≥96.0 to ≤104.0% (based on elemental C and H)
Elemental Analysis
C: 63.95-78.17% (Theory: 71.06%)
Elemental Analysis
H: 9.01-11.01% (Theory: 10.01%)

Description

Methyl cholate, is used as a bile acid derivative as anti-inflammatory and antitumor agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl cholate, is used as a bile acid derivative as anti-inflammatory and antitumor agent.

Solubility
Insoluble in water. Soluble in medium polarity organic solvents.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text