Thermo Scientific Chemicals

Glyoxime, 98+%, moistened with ca 20% water, Thermo Scientific Chemicals

Catalog number: L05130.09
10 g, Each
Thermo Scientific Chemicals

Glyoxime, 98+%, moistened with ca 20% water, Thermo Scientific Chemicals

Catalog number: L05130.09
10 g, Each
Quantity
Catalog number: L05130.09
also known as L05130-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
557-30-2
IUPAC Name
N-[(1E)-2-nitrosoethenyl]hydroxylamine
Molecular Formula
C2H4N2O2
InChI Key
RUFIRPVAEJIIIS-OWOJBTEDSA-N
SMILES
ONC=CN=O
Appearance (Color)
White to cream to pale pink or pale brown
Assay (HPLC)
≥98.0%
Form
Crystals or powder or crystalline powder or lumps or granules
Water Content (Karl Fischer Titration)
17-23%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text