9-Aminofluorene hydrochloride, 98+%, Thermo Scientific Chemicals
9-Aminofluorene hydrochloride, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

9-Aminofluorene hydrochloride, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L05156.03
also known as L05156-03
Price (USD)/ Each
25.20
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
25.20
Add to cart
9-Aminofluorene hydrochloride, 98+%, Thermo Scientific Chemicals
Catalog numberL05156.03
Price (USD)/ Each
25.20
-
Add to cart
Chemical Identifiers
CAS5978-75-6
IUPAC Namehydrogen 9H-fluoren-9-amine chloride
Molecular FormulaC13H12ClN
InChI KeySYKJOJSYQSVNOM-UHFFFAOYSA-N
SMILES[H+].[Cl-].NC1C2=CC=CC=C2C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream to cream to yellow to gray
Assay (Titration ex Chloride)≥98.0 to ≤102.0%
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only