Thermo Scientific Chemicals

Diethyl 4-methoxybenzylphosphonate, 98+%, Thermo Scientific Chemicals

Catalog number: L05226.09
10 g, Each
Thermo Scientific Chemicals

Diethyl 4-methoxybenzylphosphonate, 98+%, Thermo Scientific Chemicals

Catalog number: L05226.09
10 g, Each
Quantity
Catalog number: L05226.09
also known as L05226-09
Price (USD)
Quantity
-
Have Questions?

Description

Diethyl 4-methoxybenzylphosphonate is used in synthesis of C-aryl glycosides. It acts as a reactant for synthesis of γ -Monofluorinated goniothalamin analogs via ring-opening hydrofluorination, stilbenoid derivatives with neuroprotective activity, resveratrol-chroman hybrids with antioxidant activity, Oligostilbenoids using regioselective cyclodehydration and arylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl 4-methoxybenzylphosphonate is used in synthesis of C-aryl glycosides. It acts as a reactant for synthesis of γ -Monofluorinated goniothalamin analogs via ring-opening hydrofluorination, stilbenoid derivatives with neuroprotective activity, resveratrol-chroman hybrids with antioxidant activity, Oligostilbenoids using regioselective cyclodehydration and arylation.

Solubility
Not miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text