3,4-Dichlorobenzyl bromide is used as a pharmaceutical intermediate. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,4-Dichlorobenzyl bromide is used as a pharmaceutical intermediate. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Insoluble in water.
Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, bases.
RUO – Research Use Only
RL Bai.; CM Lin.; NN Yen.; TY Liu.; E Hamel. Identification of the cysteine residue of .beta.-tubulin alkylated by the antimitotic agent 2,4-dichlorobenzyl thiocyanate, facilitated by separatio of the protein subunits of tubulin by hydrophobic column chromatography. Biochemistry. 1989, 28,(13), 5606-5612.
C Yan.; M Han.; H Wan.; G Guan. QSAR correlation of the melting points for imidazolium bromides and imidazolium chlorides ionic liquids. Fluid Phase Equilibria. 2010, 292,(1-2), 104-109.