Diethyl 1-naphthylmethylphosphonate, 90+%, Thermo Scientific Chemicals
Diethyl 1-naphthylmethylphosphonate, 90+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethyl 1-naphthylmethylphosphonate, 90+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number L05286.18
also known as L05286-18
Price (USD)/ Each
407.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
407.00
Add to cart
Diethyl 1-naphthylmethylphosphonate, 90+%, Thermo Scientific Chemicals
Catalog numberL05286.18
Price (USD)/ Each
407.00
-
Add to cart
Chemical Identifiers
CAS53575-08-9
IUPAC Namediethyl [(naphthalen-1-yl)methyl]phosphonate
Molecular FormulaC15H19O3P
InChI KeyAFNZPMQIKMSUTD-UHFFFAOYSA-N
SMILESCCOP(=O)(CC1=C2C=CC=CC2=CC=C1)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormViscous liquid
Assay (GC)> 90.0%
Refractive Index1.5565 - 1.5655 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only