Benzyldimethylsilane, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Benzyldimethylsilane, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Insoluble in water.
Notes
Store in cool
RUO – Research Use Only
General References:
- BM Trost.; ZT Ball.; T Jöge. Regioselective hydrosilylation of propargylic alcohols: an aldol surrogate. Angewandte Chemie. 2003, 115,(29), 3537-3540.
- RJ Fessenden.; MD Coon. Silicon-Substituted Medicinal Agents. Phenyl-Substituted Silacarbamates1. Journal of medicinal chemistry. 1966, 9,(2), 262-263.