1-(4-Chlorophenyl)cyclohexane-1-carboxylic acid, 95%, Thermo Scientific Chemicals
1-(4-Chlorophenyl)cyclohexane-1-carboxylic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-(4-Chlorophenyl)cyclohexane-1-carboxylic acid, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L05336.06
also known as L05336-06
Price (USD)/ Each
64.50
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
64.50
Add to cart
1-(4-Chlorophenyl)cyclohexane-1-carboxylic acid, 95%, Thermo Scientific Chemicals
Catalog numberL05336.06
Price (USD)/ Each
64.50
-
Add to cart
Chemical Identifiers
CAS58880-37-8
IUPAC Name1-(4-chlorophenyl)cyclohexane-1-carboxylate
Molecular FormulaC13H14ClO2
InChI KeyUPNXUJXIIZGXLQ-UHFFFAOYSA-M
SMILES[O-]C(=O)C1(CCCCC1)C1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥94.0 to ≤106.0%
Appearance (Color)White to grey or purple
FormPowder or granules
Identification (FTIR)Conforms
Melting Point (clear melt)146.5-158.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only