Thermo Scientific Chemicals

(+/-)-3-Methyl-2-butanol, 98%, Thermo Scientific Chemicals

Catalog number: L05347.22
100 g, Each
Thermo Scientific Chemicals

(+/-)-3-Methyl-2-butanol, 98%, Thermo Scientific Chemicals

Catalog number: L05347.22
100 g, Each
Quantity
Catalog number: L05347.22
also known as L05347-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
598-75-4
IUPAC Name
3-methylbutan-2-ol
Molecular Formula
C5H12O
InChI Key
MXLMTQWGSQIYOW-UHFFFAOYNA-N
SMILES
CC(C)C(C)O
Refractive Index
1.4080-1.4120 @ 20?C
Assay (GC)
≥97.5%
Appearance (Color)
Clear colorless
Form
Liquid

Description

3-Methyl-2-butanol has been used as a reference for microbially produced, volatile organic compounds (MVOC) in GC-MS analyses. It acts as a plant metabolite and polar solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-2-butanol has been used as a reference for microbially produced, volatile organic compounds (MVOC) in GC-MS analyses. It acts as a plant metabolite and polar solvent.

Solubility
Miscible in Ether, Alcohols. Slightly soluble in water. (2.8g/100mL, 30°C)

Notes
Incompatible materials are Strong oxidizing agents, Reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text