6-Methoxy-2(3H)-benzoxazolone, 98+%, Thermo Scientific Chemicals
6-Methoxy-2(3H)-benzoxazolone, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Methoxy-2(3H)-benzoxazolone, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number L05349.MD
also known as L05349-MD
Price (USD)/ Each
56.70
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
56.70
Add to cart
6-Methoxy-2(3H)-benzoxazolone, 98+%, Thermo Scientific Chemicals
Catalog numberL05349.MD
Price (USD)/ Each
56.70
-
Add to cart
Chemical Identifiers
CAS532-91-2
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥98.0%
FormCrystals or powder or crystalline powder
Appearance (Color)White to pale cream or pink
Melting Point (clear melt)151.0-158.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only